Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSRC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559054
Description
Immunogen sequence: KKEGDKSQSA EIWEKLNFGN KDQNVKFRKL MGIKSEDEAG CSSVDEESYK TLKQQEEVFR NLDAQYEMAR SQTHTQRGMG LGFT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
In vitro study revealed that RSRC2 might play a role in cell proliferation. RSRC2 may be a novel tumor suppressor of esophageal cancer cell growth.
Specifications
RSRC2 | |
Polyclonal | |
Unconjugated | |
RSRC2 | |
1500011J06Rik; arginine and serine rich coiled-coil 2; arginine/serine-rich coiled-coil 2; arginine/serine-rich coiled-coil protein 2; arginine/serine-rich coiled-coil protein 2-like protein; FLJ11021; Rsrc2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
65117 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L4I2 | |
RSRC2 | |
Recombinant protein corresponding to Human RSRC2. Recombinant protein control fragment (Product #RP-97682). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction