Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSRC2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987042
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7042 Supplier Bioss Supplier No. BS6097R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In vitro study revealed that RSRC2 might play a role in cell proliferation. RSRC2 may be a novel tumor suppressor of esophageal cancer cell growth.

Specifications

Antigen RSRC2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RSRC2
Gene Accession No. Q7L4I2
Gene Alias 1500011J06Rik; arginine and serine rich coiled-coil 2; arginine/serine-rich coiled-coil 2; Arginine/serine-rich coiled-coil protein 2; BOS_16553; FLJ11021; LOC100346584; Rsrc2
Gene Symbols RSRC2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RSRC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65117
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.