Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSRP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563639
Description
Immunogen sequence: TNAAKALGTT NIDLPASLRT VPSAKETSRG IGVSSNGAKP ELSEKVTEDG TR Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 63%.
RSRP1 is a protein coding gene.
Specifications
RSRP1 | |
Polyclonal | |
Unconjugated | |
RSRP1 | |
2700043I21Rik; arginine and serine rich protein 1; arginine/serine rich protein 1; arginine/serine-rich protein 1; C1orf63; D4Ucla2; D4Wsu53e; DJ465N24.2.1; HT033; MNCb-0169; NPD014; RGD1359529; RP3-465N24.4; Rsrp1; UPF0471 protein C1orf63; UPF0471 protein C1orf63 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUV0 | |
RSRP1 | |
Recombinant protein corresponding to Human RSRP1. Recombinant protein control fragment (Product #RP-101925). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction