Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531656
Description
Recommended positive controls: NT2D1. Predicted reactivity: Human (99%), Chimpanzee (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RSRP1 is a protein coding gene.
Specifications
RSRP1 | |
Polyclonal | |
Unconjugated | |
RSRP1 | |
2700043I21Rik; arginine and serine rich protein 1; arginine/serine rich protein 1; arginine/serine-rich protein 1; C1orf63; D4Ucla2; D4Wsu53e; DJ465N24.2.1; HT033; MNCb-0169; NPD014; RGD1359529; RP3-465N24.4; Rsrp1; UPF0471 protein C1orf63; UPF0471 protein C1orf63 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BUV0 | |
RSRP1 | |
Recombinant protein encompassing a sequence within the center region of human NPD014. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction