Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555270 Supplier Invitrogen™ Supplier No. PA555270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLTYNNLSEN SLPGNFFYLT TLRALYLSDN DFEILPPDIG KLTKLQILSL RDNDLISLPK EIGELTQLKE LHIQGNRLTV LPPELGNLDL TGQKQVFKAE NNPWVTPIAD QFQLGVSHVF EYIRSETYKY LYGRHMQANP EPPKKN Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 96%.

RSU1 is a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the protein was initially isolated based on its ability to inhibit v-Ras transformation. This gene encodes a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the encoded protein was initially isolated based on its ability to inhibit v-Ras transformation. Multiple alternatively spliced transcript variants for this gene have been reported; one of these variants was found only in glioma tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSU1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSU1
Gene Accession No. Q01730, Q15404
Gene Alias cubilin; ras suppressor protein 1; etID12695; FLJ31034; LOC553276 protein; ras suppressor protein 1; ras suppressor protein 1 isoform 1; ras suppressor protein 1 variant 1; ras suppressor protein 1 variant 2; ras suppressor protein 1 variant 3; ras suppressor protein 1 variant 5; Ras suppressor protein 1-like protein; RSP1; RSP-1; rsu1; Rsu-1; RsuI; si:dz63m2.1; Unknown (protein for MGC:126965)
Gene Symbols RSU1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RSU1. Recombinant protein control fragment (Product #RP-89461).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20163, 6251, 680419
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.