Learn More
RSU1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305423AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
RSU1 is a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the protein was initially isolated based on its ability to inhibit v-Ras transformation.This gene encodes a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the encoded protein was initially isolated based on its ability to inhibit v-Ras transformation. Multiple alternatively spliced transcript variants for this gene have been reported; one of these variants was found only in glioma tumors.Specifications
RSU1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q01730, Q15404 | |
RSU1 | |
Between 227 and 277 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RSU1 | |
Ras suppressor protein 1, FLJ31034, RSP-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20163, 6251 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.