Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSV/BRSV Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | RSV/BRSV |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10093586K
![]() |
Novus Biologicals
NB10093586SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10093586
![]() |
Novus Biologicals
NB10093586 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
RSV/BRSV Polyclonal specifically detects RSV/BRSV in Virus samples. It is validated for Western Blot.Specifications
RSV/BRSV | |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Virus | |
Bovine Respiratory Syncytial Virus BRSV, Bovine RSV | |
IgG | |
Affinity Purified | |
Reacts with Human Respiratory Syncytial Virus (RSV) and Bovine Respiratory Syncytial Virus (BRSV) Fusion protein of all strains of RSV BRSV in human and bovine samples. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
Residues 483-488 [FPSDEFC] of the 63 kDa RSV and BRSV F protein. This sequence represents a neutralizing epitope and is 100% conserved among all known species. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title