Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSV Fusion Protein Antibody (RSHZ19 (Felvizumab)), Alexa Fluor™ 647, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275190AF647
Description
RSV Fusion Protein Monoclonal specifically detects RSV Fusion Protein in Viral samples. It is validated for ELISA.Specifications
RSV Fusion Protein | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
RSHZ19 (Felvizumab) | |
ELISA | |
Human | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction