Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV Fusion Protein Monoclonal Antibody (10B6), Invitrogen™

Catalog No. PIMA183437
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183437 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183437 Supplier Thermo Scientific Supplier No. MA183437

Mouse Monoclonal Antibody

Mouse anti Respiratory Syncitial Virus Fusion Protein antibody, clone 9C5 recognizes the Respiratory Syncytial Virus (RSV) Fusion protein, also known as F-protein.

Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. The 63 kD RSV fusion protein of the RSS 2 strain (subtype A) directs fusion of viral and cellular membranes, resulting in viral penetration, and can direct fusion of infected cells with adjoining cells, resulting in the formation of syncytia or multi nucleated giant cells. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV Fusion Protein
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 10B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.2
Gene Alias human respiratory syncytial virus; RSV; RSV Fusion protein; Rsvgp08
Host Species Mouse
Immunogen Purified Respiratory Syncytial Virus (RSV)
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.