Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSV Fusion Protein Recombinant Rabbit Monoclonal Antibody (R338)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA543121
This item is not returnable.
View return policy
Description
Specificity: Human respiratory syncytial virus (RSV) Fusion glycoprotein/RSV-F.
Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. The 63 kD RSV fusion protein of the RSS 2 strain (subtype A) directs fusion of viral and cellular membranes, resulting in viral penetration, and can direct fusion of infected cells with adjoining cells, resulting in the formation of syncytia or multi nucleated giant cells. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Specifications
RSV Fusion Protein | |
Recombinant Monoclonal | |
3.5 mg/mL | |
Arginine, histidine with 120mM NaCl, 0.02% Tween 80 and no preservative; pH 6.0 | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Neutralization, Western Blot | |
R338 | |
Unconjugated | |
human respiratory syncytial virus; RSV; RSV Fusion protein; Rsvgp08 | |
Recombinant Human respiratory syncytial virus (RSV) Fusion glycoprotein/RSV-F protein. | |
200 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction