Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV M2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522828
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522828 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522828 Supplier Invitrogen™ Supplier No. PA522828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specificity: Human Respiratory Syncytial Virus (RSV) M2 protein. Immunogen sequence: LESYIGSINN ITKQSA.

The RSV (Respiratory Syncytial Virus) M2 gene encodes for two critical proteins, M2-1 and M2-2. The M2-1 protein is essential for viral transcription, acting as an elongation factor, ensuring the synthesis of full-length viral mRNAs. It associates non-covalently with the viral RNA-dependent RNA polymerase complex. Transcription regulation is influenced by the phosphorylation state of M2-1, mediated by cellular protein phosphatase 1 (PP1). In contrast, the M2-2 protein plays a significant role in regulating the balance between viral transcription and replication. These functions underscore the crucial roles of M2 gene products in the RSV lifecycle and their importance as potential targets for antiviral strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV M2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias human respiratory syncytial virus; RSV; RSV M2
Host Species Rabbit
Immunogen Reacts with residues 81-95 of the human Respiratory Syncytial Virus M2 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.