Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSV Monoclonal Antibody (3098)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516299
Description
RSV Monoclonal Antibody for ICC/IF
Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Specifications
RSV | |
Monoclonal | |
6.00 mg/mL | |
PBS with 15 mM sodium azide; pH 7.5 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
Immunocytochemistry | |
3098 | |
Unconjugated | |
human respiratory syncytial virus; RSV | |
Respiratory Syncytial virus. | |
2 mg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction