Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV Monoclonal Antibody (RSV 3132)
GREENER_CHOICE

Catalog No. PIMA516283
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516283 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516283 Supplier Invitrogen™ Supplier No. MA516283
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RSV Monoclonal Antibody for ICC/IF

Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV, Long, Randall, RSV Strains 127, SNK & 9007
Applications Immunocytochemistry
Classification Monoclonal
Clone RSV 3132
Concentration 6.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 15 mM sodium azide; pH 7.5
Gene Alias human respiratory syncytial virus; RSV
Host Species Mouse
Immunogen RSV strains 127, SNK & 9007, Long, Randall, 8/60 & A/2.
Purification Method Protein A
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.