Learn More
Invitrogen™ RSV Monoclonal Antibody (RSV 3216)

Description
RSV Monoclonal Antibody for ICC/IF
Specifications
Specifications
| Antigen | RSV, Long, Randall, RSV Strains 127, SNK & 9007 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | RSV 3216 |
| Concentration | 7.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.5 |
| Gene Alias | human respiratory syncytial virus; RSV |
| Host Species | Mouse |
| Immunogen | RSV strains 127, SNK & 9007, Long, Randall, 8/60 & A/2. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.