Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV NP Monoclonal Antibody (B023)
GREENER_CHOICE

Catalog No. MA180039
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA180039 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA180039 Supplier Invitrogen™ Supplier No. MA180039
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suitable for use as detection reagent with MA1-80661 as capture reagent in sandwich ELISA. Mouse anti Respiratory Syncytial Virus Nucleoprotein antibody, clone RSV3132 (B023) recognizes an epitope within the 42kD RSV nucleoprotein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV NP
Applications ELISA, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone B023
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Gene Alias human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein
Host Species Mouse
Immunogen Bovine RSV strains: 127, SNK and 9007. Human RSV strains: A2, Long, Randall and 8/60.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.