Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSV NP Recombinant Rabbit Monoclonal Antibody (HL1247)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA546973
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.

Specifications
RSV NP | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Immunoprecipitation, Lateral Flow, Western Blot | |
HL1247 | |
Unconjugated | |
human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein | |
Recombinant protein encompassing a sequence within the C-terminus region of Respiratory Syncytial virus Nucleoprotein. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction