Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV NP Monoclonal Antibody (I25J), FITC
GREENER_CHOICE

Catalog No. PIMA17289
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17289 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17289 Supplier Invitrogen™ Supplier No. MA17289
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7289 antibody reacts with Respiratory Sync. virus Np in viral and infected tissue samples. MA1-7289 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7289 antibody was raised against RSV nucleoprotein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV NP
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone I25J
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein
Host Species Mouse
Immunogen RSV nucleoprotein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.