Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV NP Recombinant Rabbit Monoclonal Antibody (HL1296)
GREENER_CHOICE

Catalog No. FEMA550310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550310 Supplier Invitrogen™ Supplier No. MA550310
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product can be used in Sandwich ELISA to detect RSV NP using MA5-46972 as a capture antibody and MA5-50310 as the detection antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV NP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1296
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Respiratory Syncytial virus Nucleoprotein. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.