Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV NS1 Recombinant Rabbit Monoclonal Antibody (HL2383)
GREENER_CHOICE

Catalog No. FEMA550511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550511 Supplier Invitrogen™ Supplier No. MA550511
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

RSV NS1 Recombinant Monoclonal Antibody for Western Blot

RSV NS1 is a gene target that is found in respiratory syncytial virus (RSV), a common cause of respiratory infections in infants and young children. NS1 is a non-structural protein that plays a key role in the replication of the virus, and is therefore a potential target for antiviral drugs. The NS1 protein interacts with host cell proteins and interferes with the host immune response, making it difficult for the body to fight off the virus. Studies have shown that targeting NS1 can reduce viral replication and improve outcomes in animal models of RSV infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV NS1
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL2383
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Respiratory Syncytial virus NS1
Host Species Rabbit
Immunogen Recombinant fragment of Respiratory Syncytial virus NS1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.