Learn More
Invitrogen™ RSV Polyclonal Antibody, Biotin

Description
The PA1-73019 antibody reacts with Respiratory Syncytial virus. PA1-73019 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73019 antibody was raised against Human RSV isolate (confirmed).
Specifications
Specifications
| Antigen | RSV |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | human respiratory syncytial virus; RSV |
| Host Species | Goat |
| Immunogen | Human RSV isolate (confirmed). |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.