Learn More
Invitrogen™ RSV Polyclonal Antibody, HRP

Description
The PA1-73018 antibody reacts with Respiratory Syncytial virus. PA1-73018 has been successfully used in ELISA applications. The PA1-73018 antibody was raised against Human RSV isolate (confirmed). Store at 4°C for up to 6 months. For extended storage, store at -20°C, avoiding freeze/thaw cycles.
Specifications
Specifications
| Antigen | RSV |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 10mg/mL BSA and 0.002% thimerosal |
| Gene Alias | human respiratory syncytial virus; RSV |
| Host Species | Goat |
| Immunogen | Human RSV isolate (confirmed). |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.