Learn More
Invitrogen™ RSV strain A2 Polyclonal Antibody

Description
PA1-4181 detects Respiratory Syncytial Virus (A2 Strain). PA1-4181 has been successfully used in Western blot procedures. The PA1-4181 immunogen is a synthetic peptide corresponding to residues T(180) C W A I C K R I P N K K P G K K(198) of RSV G (A2 strain).
Specifications
Specifications
| Antigen | RSV strain A2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | human respiratory syncytial virus; RSV; RSV A2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to residues T(180) C W A I C K R I P N K K P G K K(198) of human RSV G (A2 strain). |
| Purification Method | Antigen Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.