Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV strain A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522827
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522827 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522827 Supplier Invitrogen™ Supplier No. PA522827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RSV strain A2 Polyclonal Antibody for Western Blot

Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV strain A2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias human respiratory syncytial virus; RSV; RSV A2
Host Species Rabbit
Immunogen Reacts with residues 180-198 of the human RSV G protein A2 strain.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.