Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV strain B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14182
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14182 Supplier Invitrogen™ Supplier No. PA14182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4182 detects Respiratory Syncytial Virus (B1 Strain). PA1-4182 has been successfully used in Western blot procedures. The PA1-4182 immunogen is a synthetic peptide corresponding to residues L(187) C K S I C K T I P S N K P K K K P(198) of RSV G (B1 strain).

Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV strain B1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias human respiratory syncytial virus; RSV; RSV B1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues L(187) C K S I C K T I P S N K P K K K P(198) of RSV G (B1 strain).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.