Learn More
Invitrogen™ RSV strain RSS-2 Polyclonal Antibody

Description
RSV strain RSS-2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | RSV strain RSS-2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | human respiratory syncytial virus; respiratory syncytial virus strain rss-2; RSV; RSV RSS-2 |
| Host Species | Rabbit |
| Immunogen | Reacts with residues 201-211 of the 63 kDa RSV F protein. |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.