Learn More
Invitrogen™ RSV strain RSS-2 Polyclonal Antibody

Description
PA1-4102 detects the respiratory syncytial virus strain RSS-2. PA1-4102 has been successfully used in Western blot procedures. The PA1-4102 immunogen is hRSV fusion protein of RSS-2 (subtype A) strain. This antibody reacts with residues K(201) Q L L P I V N K Q S(211) C of RSV F. Maintain at 4ºC for 3 months or at -20ºC for longer periods. Stable for 1 year. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | RSV strain RSS-2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | human respiratory syncytial virus; respiratory syncytial virus strain rss-2; RSV; RSV RSS-2 |
| Host Species | Rabbit |
| Immunogen | hRSV fusion protein of RSS-2 (subtype A) strain. |
| Purification Method | Antigen Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.