Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RSV Type A NP Recombinant Rabbit Monoclonal Antibody (HL1297)
GREENER_CHOICE

Catalog No. PIMA546992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546992 Supplier Invitrogen™ Supplier No. MA546992
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Respiratory syncytial virus (RSV) is an important human pathogen Its nucleocapsid (NC), which comprises the negative sense RNA viral genome coated by the viral nucleoprotein N, is a critical assembly that serves as template for both mRNA synthesis and genome replication [PMID: 23677789] RSV Type A is more prevalent than type B and seems to be the cause of the most severe cases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV Type A NP
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1297
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Respiratory Syncytial virus Nucleoprotein. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.