Learn More
Description
Includes
All necessary solutions, buffers, chemicals and templates for six sets of reactions; complete instructions; background information and study questions
Requires
- Horizontal gel electrophoresis apparatus
- Thermal cycler
- Hotplate
- Micropipetter
- Balance
- Water bath
- 250mL Flasks
- UV transilluminator
- Pipet pump or bulb
- All necessary safety equipment
- Distilled or deionized water
Similar to HIV replication
- Reactions of the provided mRNA are reverse transcribed to double-stranded DNA
- Amplify the DNA using PCR in order to see the results of this reaction
Order Info
Supplies for six reactions
Specifications
Specifications
| Content And Storage | Instructions, RNA template, primer mix, RT-PCR reaction beads, RNase free water, DNA size ladder, Agarose, electrophoresis Buffer, InstaStain™ Ethidium Bromide. Store at -20°C |
| Detection Method | Gel Electrophoresis |
| Form | Lyophilized |
| GC-Rich PCR Performance | Medium |
| PCR Method | RT-PCR |
| Reaction Speed | 30 to 90 min. |
| Kit Type | RT-PCR kit |
| Label or Dye | SYBR Green, FlashBlue Stain |
| Product Type | Classroom Experiment |
| For Use With (Application) | RT-PCR |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

