Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class I (RT1-A) Monoclonal Antibody (MRC OX-18)
GREENER_CHOICE

Catalog No. PIMA190799
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA190799 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA190799 Supplier Invitrogen™ Supplier No. MA190799
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-90799 detects RT1a Class I from rat samples.

This protein is a non-polymorphic determinant of rat MHC Class I, which is also known as RT1-A. Rat MHC Class I is expressed on all nucleated cells and erythrocytes. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class I (RT1-A)
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone MRC OX-18
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RT1-A
Gene Accession No. 0
Gene Alias HA11; MHC class I RT1.Aa alpha-chain; Rat class I A; RT1 class I gene (A-1); RT1 class Ia, locus A (A1); RT1 class Ia, locus A1; RT1.A1(N); Rt1.aa; RT1-A; RT1-A1; RT1A-1; RT1-AW2
Gene Symbols RT1-A, RT1-A1
Host Species Mouse
Immunogen Rat spleen cell glycoprotein.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100188935, 24973
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.