Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RT1.Ac Monoclonal Antibody (OX-27), Biotin
RT1.Ac Monoclonal Antibody (OX-27), Biotin
Supplier: Thermo Scientific MA517464
Description

Specifications
RT1.Ac | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen) | |
OX-27 | |
Biotin | |
RT1-Ac | |
Phytohaemagglutinin Blasts. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction