Learn More
Invitrogen™ RT1.B Monoclonal Antibody (OX-6)
Mouse Monoclonal Antibody
$355.00 - $1052.00
Specifications
Antigen | RT1.B |
---|---|
Clone | OX-6 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA180672
|
Invitrogen™
MA180672 |
1 mg |
Each for $1,052.00
|
|
|||||
MA181523
|
Invitrogen™
MA181523 |
100 μg |
Each for $355.00
|
|
|||||
Description
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. PLP fixation is recommended for optimal results. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat MHC Class II RT1B antibody, clone OX-6 recognizes a monomorphic determinant of the rat RT1B MHC class II antigen present on B lymphocytes, dendritic cells, some macrophages and certain epithelial cells. Mouse anti Rat MHC Class II RT1B antibody, clone OX-6 recognizes a monomorphic determinant of the rat RT1B MHC class II antigen present on B lymphocytes, dendritic cells, some macrophages and certain epithelial cells.
gene complex that may play a role in antigen presentation.Specifications
RT1.B | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
0 | |
24738 | |
Rat thymocyte membrane glycoprotein. | |
Antibody |
OX-6 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG1 | |
Mouse, Rat | |
RT1-B | |
rat class I B; RT1 class II, locus B; RT1-B | |
RT1-B | |
Primary | |
Protein G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.