Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RT1.B Monoclonal Antibody (OX-6), FITC
GREENER_CHOICE

Catalog No. PIMA517430
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517430 100 μg
PIMA517431 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517430 Supplier Invitrogen™ Supplier No. MA517430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RT1.B Monoclonal Antibody for Flow

gene complex that may play a role in antigen presentation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RT1.B
Applications Flow Cytometry
Classification Monoclonal
Clone OX-6
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene RT1-B
Gene Accession No. 0
Gene Alias rat class I B; RT1 class II, locus B; RT1-B
Gene Symbols RT1-B
Host Species Mouse
Immunogen rat thymocyte membrane glycoprotein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24738
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.