Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RT1.B Monoclonal Antibody (OX-6), PE
Mouse Monoclonal Antibody
$318.00 - $633.50
Specifications
Antigen | RT1.B |
---|---|
Clone | OX-6 |
Concentration | 0.1 mg/mL |
Content And Storage | 4°C, do not freeze |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517432
|
Invitrogen™
MA517432 |
50 μg |
Each for $318.00
|
|
|||||
PIMA517433
|
Invitrogen™
MA517433 |
200 μg |
Each for $633.50
|
|
|||||
Description
RT1.B Monoclonal Antibody for Flow
gene complex that may play a role in antigen presentation.Specifications
RT1.B | |
0.1 mg/mL | |
Flow Cytometry | |
PE | |
Mouse | |
RUO | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
0 | |
24738 | |
rat thymocyte membrane glycoprotein. | |
Antibody |
OX-6 | |
4°C, do not freeze | |
Monoclonal | |
Liquid | |
IgG1 | |
Rat | |
RT1-B | |
rat class I B; RT1 class II, locus B; RT1-B | |
RT1-B | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title