Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RT1.Bu Monoclonal Antibody (OX-3), PE

Catalog No. PIMA517419
Change view
Click to view available options
Quantity:
200 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517419 200 μg
PIMA517418 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517419 Supplier Invitrogen™ Supplier No. MA517419
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

anti-rat RT1.Bu monoclonal antibody recognizes a polymorphic determinant on rat Ia present on Lewis, Wistar, and AO rat strains (although it is apparently expressed in lower amounts in AO rats) but not BN, DA, or PVG/c strains. This antibody also cross-reacts with mouse strains (MHC haplotypes b and s) and analysis of recombinant mouse strains shows that the determinant maps to the I-A region and correlates with mouse Ia specificity 9. Recognizes Ia antigens on approximately 18% of thymocytes but does not significantly label the majority of peripheral T lymphocytes. Thus, this antibody is particularly useful for distinguishing Ia positive cells from different rat strains and has been used in the recognition of cells of donor origin in bone marrow reconstituted radiation chimeras.

RT1.Bu is a gene located within the major histocompatibility complex (MHC) region of the rat genome, specifically part of the RT1 complex. The RT1 complex in rats is analogous to the human HLA and mouse H2 MHC regions and encompasses a variety of class I and class II genes involved in antigen presentation. RT1.Bu is one such class I MHC gene, encoding a protein essential for the presentation of endogenous peptides to cytotoxic T cells. This interaction is vital for the immune system's ability to recognize and eliminate infected or malignantly transformed cells. The RT1.Bu gene is crucial for studying immune responses, particularly in transplantation immunology and autoimmune disease models, due to its role in histocompatibility and immune regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RT1.Bu
Applications Flow Cytometry
Classification Monoclonal
Clone OX-3
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene RT1-Ba
Gene Accession No. P20037
Gene Alias activated B cell RT1Bl alpha chain; BA1; MHC class II antigen RT1.B alpha chain; rano class II histocompatibility antigen, B alpha chain; rat class I Bu; RT1 class II antigen, Ba chain; RT1 class II, locus Ba; RT1-B alpha; RT1-Ba; RT1-Bu
Gene Symbols RT1-Ba
Host Species Mouse
Immunogen Rat thymocyte membrane glycoprotein.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309621
Target Species Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.