Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RT1.D Monoclonal Antibody (OX-17), Biotin
GREENER_CHOICE

Catalog No. PIMA517435
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517435 100 μg
PIMA517436 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517435 Supplier Invitrogen™ Supplier No. MA517435
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

anti-rat RT1.D monoclonal antibody recognizes a monomorphic determinant on the α chain of the rat Ia antigen and appears to be the rat homologue of mouse Ia-E. It recognizes the rat Ia product present on B, but not T cells from lymph node or thoracic duct lymph. It does not bind to thymocytes or erythrocytes. The antibody does not cross-react with rat Ia-A or mouse Ia-E antigen, but rabbit antibody raised against the antibody affinity column-purified MRC OX-17 antigen cross-reacted on tissues of mice expressing Ia-E mouse antigen but not on those mouse strains that were Ia-E antigen negative.

RT1.D is a gene located within the major histocompatibility complex (MHC) region of the rat genome, specifically part of the RT1 complex. RT1.D, along with its counterpart RT1.B, comprises class II MHC genes, which are key players in the adaptive immune response. These genes encode for proteins that present extracellularly derived peptides to helper T cells (CD4+ T cells), facilitating the activation and regulation of immune responses. The RT1.D gene product is analogous to human HLA-DR and mouse I-A/I-E molecules, playing an essential role in antigen presentation and immune recognition. RT1.D genes are highly polymorphic, which influences susceptibility to various diseases and are often utilized in studies involving autoimmune diseases, immune response, and transplantation immunology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RT1.D
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX-17
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene RT1-Db1
Gene Accession No. P18211
Gene Alias Db1; MHC class II RT1D beta1 chain antigen; rano class II histocompatibility antigen, D-1 beta chain; rat class I D; RT1 class II histocompatibility antigen, D-1 beta chain; RT1 class II, locus Db1; RT1-D beta; RT1-Db; RT1-Db1; RT1-Db1n
Gene Symbols RT1-Db1
Host Species Mouse
Immunogen Rat spleen membrane glycoproteins depleted of Ia-A antigens.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294270
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.