Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RT28 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555542
Description
Immunogen sequence: MAALCRTRAV AAESHFLRVF LFFRPFRGVG TESGSESGSS NAKEPKTRAG GFASALERHS ELLQKVEPLQ KGSPKNVESF ASMLRHSPLT QM Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 60%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that has been called mitochondrial ribosomal protein S35 in the literature.
Specifications
RT28 | |
Polyclonal | |
Unconjugated | |
MRPS28 | |
1500012D08Rik; 28S ribosomal protein S28, mitochondrial; 28S ribosomal protein S35, mitochondrial; AW061224; HSPC007; LRRGT00031; mitochondrial 28S ribosomal protein S35; mitochondrial ribosomal protein S28; mitochondrial ribosomal protein S28-like; Mitochondrial small ribosomal subunit protein bS1m; Mrps28; MRP-S28; Mrps28l; MRPS35; MRP-S35; S28mt; S35mt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2Q9 | |
MRPS28 | |
Recombinant protein corresponding to Human RT28. Recombinant protein control fragment (Product #RP-93813). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction