Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RT28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106942 Supplier Invitrogen™ Supplier No. PA5106942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MRPS28.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that has been called mitochondrial ribosomal protein S35 in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RT28
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MRPS28
Gene Accession No. Q9CY16, Q9Y2Q9
Gene Alias 1500012D08Rik; 28S ribosomal protein S28, mitochondrial; 28S ribosomal protein S35, mitochondrial; AW061224; HSPC007; LRRGT00031; mitochondrial 28S ribosomal protein S35; mitochondrial ribosomal protein S28; mitochondrial ribosomal protein S28-like; Mitochondrial small ribosomal subunit protein bS1m; Mrps28; MRP-S28; Mrps28l; MRPS35; MRP-S35; S28mt; S35mt
Gene Symbols MRPS28
Host Species Rabbit
Immunogen A synthesized peptide derived from human MRPS28(Accession Q9Y2Q9), corresponding to amino acid residues L154-K187.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28957, 66230, 689025
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.