Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTBDN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590530 Supplier Invitrogen™ Supplier No. PA590530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRPLQARSQQ HHGLAADLGK GKLHLAGPCC PSEMDTTETS GPGNHPERCG VPSPECESFL EHLQRALRSR FRLRLLGVRQ AQPLCEELCQ AWFANCEDDI TCGPTWLPLS EKRGCEPSCL TYGQTFADGT DLCRSALGHA LPVAAPGARH CFNISISAVP RPRPGRRGRE APSRRSRSPR TSILDAAGSG SGSGSGSGP; Positive Samples: Rat eye.

This gene was first identified in a study of human eye tissues. The protein encoded by this gene is preferentially expressed in the retina and may play a role in binding retinoids and other carotenoids as it shares homology with riboflavin binding proteins. Alternative splicing results in multiple transcript variants and protein isoforms. [provided by RefSeq, Jul 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTBDN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RTBDN
Gene Accession No. Q9BSG5
Gene Alias FLJ36353; retbindin; RTBDN; Rtbnd
Gene Symbols RTBDN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 63-261 of human RTBDN (NP_1136171).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304667, 83546
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.