Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol, Certified Reference Material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
20 mL
Specifications
Specifications
| CAS | 67-56-1 |
| Vapor Pressure | 410 mmHg (50°C) |
| Vapor Density | 1.11 (vs air) |
| Density | 0.791 g/mL (at 25°C (literature)) |
| Flash Point | 9.7°C (Not Applicable) |
| Molecular Formula | CH4O |
| Linear Formula | CH3OH |
| MDL Number | MFCD00004595 |
| Melting Point | -98°C (literature) |
| Boiling Point | 64.7°C (literature) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction