Learn More
RTCB/C22orf28/FAAP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305079AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. tropicalis, Bovine and Pig
The function of this protein remains unknown.Specifications
RTCB/C22orf28/FAAP | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q99LF4, Q9Y3I0 | |
Rtcb | |
Between 455 and 505 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RTCB | |
ankyrin repeat domain 54, C22orf28, chromosome 22 open reading frame 28, DJ149A16.6, FAAP, focal adhesion-associated protein, HSPC117 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28088, 51493 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.