Learn More
RTCB/C22orf28/FAAP Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305078A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. tropicalis, Bovine and Pig
The function of this protein remains unknown.Specifications
RTCB/C22orf28/FAAP | |
Polyclonal | |
Unconjugated | |
RTCB | |
ankyrin repeat domain 54, C22orf28, chromosome 22 open reading frame 28, DJ149A16.6, FAAP, focal adhesion-associated protein, HSPC117 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28088, 51493 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q99LF4, Q9Y3I0 | |
RTCB | |
Between 175 and 225 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.