Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544610 Supplier Invitrogen™ Supplier No. PA544610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PEAVVSPGGV GFDINCGVRL LRTNLDESDV QPVKEQLAQA MFDHIPVGVG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTCB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Rtcb
Gene Accession No. Q9Y3I0
Gene Alias 3'-phosphate/5'-hydroxy nucleic acid ligase; AI255213; AI463255; ankyrin repeat domain 54; C22orf28; C5H22orf28; D10Wsu52e; DJ149A16.6; FAAP; focal adhesion-associated protein; HAMAP-Rule:MF_03144}; HSPC117; hypothetical protein HSPC117; hypothetical protein LOC406376; hypothetical protein LOC525106; P55; RNA 2',3'-cyclic phosphate and 5'-OH ligase; RNA-splicing ligase RtcB homolog; RTCB; tRNA-splicing ligase RtcB homolog; tRNA-splicing ligase RtcB homolog {ECO:0000255; zgc:76871
Gene Symbols Rtcb
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C22orf28 (aa 107-156).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51493
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.