Learn More
Invitrogen™ RTCB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564867
Description
Immunogen sequence: PDDLDLHVIYD VSHNIAKVEQ HVVDGKERTL LVHRKGSTRA FPPHHPLIAV DYQLTGQPVL IGGTMGTCSY VLTGTEQGMT ETFGTTCHGA GRALSRAKSR RNLDFQDVLD KLADMGIAIR VASPKLVMEE APESYKNVTD VVNTCHDAGI SKKAIKLRP Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The function of this protein remains unknown.
Specifications
RTCB | |
Polyclonal | |
Unconjugated | |
Rtcb | |
3'-phosphate/5'-hydroxy nucleic acid ligase; AI255213; AI463255; ankyrin repeat domain 54; C22orf28; C5H22orf28; D10Wsu52e; DJ149A16.6; FAAP; focal adhesion-associated protein; HAMAP-Rule:MF_03144}; HSPC117; hypothetical protein HSPC117; hypothetical protein LOC406376; hypothetical protein LOC525106; P55; RNA 2',3'-cyclic phosphate and 5'-OH ligase; RNA-splicing ligase RtcB homolog; RTCB; tRNA-splicing ligase RtcB homolog; tRNA-splicing ligase RtcB homolog {ECO:0000255; zgc:76871 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51493 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3I0 | |
Rtcb | |
Recombinant Human RTCB. Recombinant protein control fragment (Product #RP-91392). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.