Learn More
Invitrogen™ RTCB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587793
Description
Immunogen sequence: EIFNEYAAKK MGIDHKGQVC VMIHSGSRGL GHQVATDALV AMEKAMKRDK IIVNDRQLAC ARIASPEGQD YLKGMAAAGN YAWVNRSSMT FLTRQAFAKV FNTTPDDLDL HVIYDVSHNI AKVEQHVVDG KERTLLVHRK GSTRAFPPHH PLIAVDYQLT GQPVLIGGTM GTCSYVLTGT EQGMTETFGT TCHGAGRALS RAKSRRNLDF QDVLDKLADM GIAIRVASPK LVMEEAPESY KNVTDVVNTC HDAGISKKAI KLRPIAVIKG; Positive Samples: A-549, Mouse liver, Rat liver; Cellular Location: Cytoplasm, Nucleus.
The function of this protein remains unknown.
Specifications
RTCB | |
Polyclonal | |
Unconjugated | |
Rtcb | |
3'-phosphate/5'-hydroxy nucleic acid ligase; AI255213; AI463255; ankyrin repeat domain 54; C22orf28; C5H22orf28; D10Wsu52e; DJ149A16.6; FAAP; focal adhesion-associated protein; HAMAP-Rule:MF_03144}; HSPC117; hypothetical protein HSPC117; hypothetical protein LOC406376; hypothetical protein LOC525106; P55; RNA 2',3'-cyclic phosphate and 5'-OH ligase; RNA-splicing ligase RtcB homolog; RTCB; tRNA-splicing ligase RtcB homolog; tRNA-splicing ligase RtcB homolog {ECO:0000255; zgc:76871 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
28088, 362855, 51493 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AYT3, Q99LF4, Q9Y3I0 | |
Rtcb | |
Recombinant fusion protein containing a sequence corresponding to amino acids 236-505 of human RTCB (NP_0551211). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.