Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142753
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142753 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142753 Supplier Invitrogen™ Supplier No. PA5142753
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTCB
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Rtcb
Gene Accession No. Q9Y3I0
Gene Alias 3'-phosphate/5'-hydroxy nucleic acid ligase; AI255213; AI463255; ankyrin repeat domain 54; C22orf28; C5H22orf28; D10Wsu52e; DJ149A16.6; FAAP; focal adhesion-associated protein; HAMAP-Rule:MF_03144}; HSPC117; hypothetical protein HSPC117; hypothetical protein LOC406376; hypothetical protein LOC525106; P55; RNA 2',3'-cyclic phosphate and 5'-OH ligase; RNA-splicing ligase RtcB homolog; RTCB; tRNA-splicing ligase RtcB homolog; tRNA-splicing ligase RtcB homolog {ECO:0000255; zgc:76871
Gene Symbols Rtcb
Host Species Goat
Immunogen Peptide with sequence C-QADPNKVSARAKKR.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51493
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.