Learn More
Invitrogen™ RTDR1 Monoclonal Antibody (3B6)

Description
Peptide Sequence: SNAAGALMFA TVITEGKYAA LEAQAIGLLL ELLHSPMTIA RLNATKALTM LAEAPEGRKA LQTHVPTFRA MEVETYEKPQ VAEALQRAAR IAISVIEFKP.
Specifications
Specifications
| Antigen | RTDR1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3B6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RSPH14 |
| Gene Accession No. | Q9UHP6 |
| Gene Alias | 4933431K05Rik; MGC16968; Radial spoke head 14 homolog; radial spoke head homolog 14 (Chlamydomonas); rhabdoid tumor deletion region gene 1; rhabdoid tumor deletion region protein 1; Rsph14; Rtdr1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.