Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTDR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553796 Supplier Invitrogen™ Supplier No. PA553796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLELPININA TQITTAYGHR ALPKLKEELQ SEDLQTRQKA LMALCDLMHD PECIYKAMNI GCMENLKALL KD Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 69%.

RTDR1 (Rhabdoid tumor deletion region protein 1) is a 348 amino acid protein that is expressed in adult spinal cord, skeletal muscle, cerebellum and spleen, as well in some rhabdoid tumors. The gene encoding RTDR1 maps to human chromosome 22, which contains over 500 genes and about 49 million bases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTDR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSPH14
Gene Accession No. Q9UHP6
Gene Alias 4933431K05Rik; MGC16968; Radial spoke head 14 homolog; radial spoke head homolog 14 (Chlamydomonas); rhabdoid tumor deletion region gene 1; rhabdoid tumor deletion region protein 1; Rsph14; Rtdr1
Gene Symbols RSPH14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RTDR1. Recombinant protein control fragment (Product #RP-92351).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27156
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.