Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTDR1 Polyclonal Antibody

Supplier: Thermo Scientific PA5113475
Description
Specifications
| RTDR1 | |
| Polyclonal | |
| Unconjugated | |
| RSPH14 | |
| 4933431K05Rik; MGC16968; Radial spoke head 14 homolog; radial spoke head homolog 14 (Chlamydomonas); rhabdoid tumor deletion region gene 1; rhabdoid tumor deletion region protein 1; Rsph14; Rtdr1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 27156 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UHP6 | |
| RSPH14 | |
| Recombinant Human Radial spoke head 14 homolog protein (1-230AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction