Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTEL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566911 Supplier Invitrogen™ Supplier No. PA566911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKKHNLLQGFY QFVRPHHKQQ FEEVCIQLTG RGCGYRPEHS IPRRQRAQPV LDPTGRTAPD PKLTVSTAAA QQLDPQEHLN QG Highest antigen sequence identity to the following orthologs - mouse 53%, rat 60%.

ATP-dependent DNA helicase implicated in telomere-length regulation, DNA repair and the maintenance of genomic stability. Acts as an anti-recombinase to counteract toxic recombination and limit crossover during meiosis. Regulates meiotic recombination and crossover homeostasis by physically dissociating strand invasion events and thereby promotes noncrossover repair by meiotic synthesis dependent strand annealing (SDSA) as well as disassembly of D loop recombination intermediates. Also disassembles T loops and prevents telomere fragility by counteracting telomeric G4-DNA structures, which together ensure the dynamics and stability of the telomere.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTEL1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rtel1
Gene Accession No. Q9NZ71
Gene Alias AI451565; AW540478; C20orf41; DEAH helicase; DKCA4; DKCB5; HAMAP-Rule:MF_03065}; helicase-like protein NHL; KIAA1088; NHL; Novel helicase-like; PFBMFT3; Regulator of telomere elongation helicase 1; regulator of telomere elongation helicase 1 {ECO:0000255; regulator of telomere length; RGD1306721; Rtel; Rtel1
Gene Symbols Rtel1
Host Species Rabbit
Immunogen Recombinant Human RTEL1. Recombinant protein control fragment (Product #RP-105423).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51750
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.