Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTF1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600277AF532
Description
RTF1 Polyclonal antibody specifically detects RTF1 in Human samples. It is validated for Western BlotSpecifications
RTF1 | |
Polyclonal | |
Western Blot | |
KIAA0252GTL7, ortholog of mouse gene trap locus 7, RNA polymerase-associated protein RTF1 homolog, Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae) | |
The immunogen recognized by this antibody maps to a region between residues 300 and 350 of human Restores TBP (TATA box-binding protein) function-1 using the numbering given in entry EAW92492.1 (GeneID 23168). | |
0.1 mL | |
Cell Cycle and Replication | |
23168 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction